missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-3-(trifluoromethyl)pyridine, 96%

Manufacturer:  Alfa Aesar™ H5430503

 View more versions of this product

Catalog No. AAH5430503



Specifications

Specifications

2-Amino-5-bromo-3-(trifluoromethyl)pyridine
79456-34-1
MFCD10758088
QUYZBNHTYCLZLW-UHFFFAOYSA-N
5-bromo-3-(trifluoromethyl)pyridin-2-amine
20525383
96%
1g
C6H4BrF3N2
2-amino-5-bromo-3-trifluoromethyl pyridine
C1=C(C(=NC=C1Br)N)C(F)(F)F
241.011
241.01

Chemical Identifiers

79456-34-1
241.011
QUYZBNHTYCLZLW-UHFFFAOYSA-N
20525383
C1=C(C(=NC=C1Br)N)C(F)(F)F
C6H4BrF3N2
MFCD10758088
2-amino-5-bromo-3-trifluoromethyl pyridine
5-bromo-3-(trifluoromethyl)pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents