Please login to your online account to display your discounted pricing

2-Amino-5-bromo-3-nitropyridine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-5-bromo-3-nitropyridine
CAS 6945-68-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H4BrN3O2
Formula Weight 218.02
MDL Number MFCD00047441
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306240010

  SDS
thermo fisher scientific
306240010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 218.02
Quantity 1g
Color Yellow-brown
Melting Point 205° to 210°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2-Amino-5-bromo-3-nitropyridine
Assay Percent Range 98%
Molecular Formula C5H4BrN3O2
MDL Number MFCD00047441
CAS 6945-68-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.