missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-3-nitropyridine, 97%

Manufacturer:  Alfa Aesar™ A1498503

 View more versions of this product

Catalog No. AAA1498503



Specifications

Specifications

2-Amino-5-bromo-3-nitropyridine
1g
C5H4BrN3O2
383851
QOOCOFOGYRQPPN-UHFFFAOYSA-N
5-bromo-3-nitropyridin-2-amine
138878
97%
206°C to 210°C
6945-68-2
MFCD00047441
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
C1=C(C(=NC=C1Br)N)[N+](=O)[O-]
218.01
218.01

Chemical Identifiers

6945-68-2
218.01
QOOCOFOGYRQPPN-UHFFFAOYSA-N
138878
C1=C(C(=NC=C1Br)N)[N+](=O)[O-]
C5H4BrN3O2
MFCD00047441
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
5-bromo-3-nitropyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents