missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-3-methylpyridine, 97%

Manufacturer:  Alfa Aesar™ A1531803

 View more versions of this product

Catalog No. AAA1531803



Specifications

Specifications

2-Amino-5-bromo-3-methylpyridine
90°C to 94°C
3430-21-5
MFCD00068232
2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
CC1=CC(=CN=C1N)Br
187.04
187.04
Unpleasant
1g
C6H7BrN2
386426
KBLGGRWUEVCNPY-UHFFFAOYSA-N
5-bromo-3-methylpyridin-2-amine
137934
97%

Chemical Identifiers

3430-21-5
187.04
KBLGGRWUEVCNPY-UHFFFAOYSA-N
137934
CC1=CC(=CN=C1N)Br
C6H7BrN2
MFCD00068232
2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
5-bromo-3-methylpyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents