missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-3-hydroxypyridine, 95%

Manufacturer:  Alfa Aesar™ H61722MD

 View more versions of this product

Catalog No. AAH61722MD



Description

Description

Applications
2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

2-Amino-5-bromo-3-hydroxypyridine
250mg
C5H5BrN2O
2-amino-3-hydroxy-5-bromo pyridine
C1=C(C(=NC=C1Br)N)O
189.012
189.01
95%
White
39903-01-0
MFCD09744143
YQADLKDQAXAIKW-UHFFFAOYSA-N
2-amino-5-bromopyridin-3-ol
11694041
Solid

Chemical Identifiers

39903-01-0
189.012
YQADLKDQAXAIKW-UHFFFAOYSA-N
11694041
C1=C(C(=NC=C1Br)N)O
C5H5BrN2O
MFCD09744143
2-amino-3-hydroxy-5-bromo pyridine
2-amino-5-bromopyridin-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents