Please login to your online account to display your discounted pricing

2-Amino-5,6-dimethylbenzothiazole 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103550250

 View more versions of this product

Catalog No. AC103550250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Melting Point >180°C
Formula Weight 178.25
Quantity 25g
Color White to yellow or beige
EINECS Number 249-960-4
CAS Max % 100.0
Name Note 98%
CAS 29927-08-0
Assay Percent Range 98%
Chemical Name or Material 2-Amino-5, 6-dimethylbenzothiazole
Molecular Formula C9 H10 N2 S
CAS Min % 97.5
MDL Number MFCD00005790
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Amino-5, 6-dimethylbenzothiazole
Name Note 98%
CAS 29927-08-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H10 N2 S
Formula Weight 178.25
MDL Number MFCD00005790