Please login to your online account to display your discounted pricing

2-Amino-5,6-dimethylbenzothiazole 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-5, 6-dimethylbenzothiazole
Name Note 98%
CAS 29927-08-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H10 N2 S
Formula Weight 178.25
MDL Number MFCD00005790
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC103550250

  SDS
thermo fisher scientific
103550250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Melting Point >180°C
Physical Form Crystalline Powder
Formula Weight 178.25
Quantity 25g
Color White to yellow or beige
EINECS Number 249-960-4
CAS Max % 100.0
Name Note 98%
CAS 29927-08-0
Chemical Name or Material 2-Amino-5, 6-dimethylbenzothiazole
Assay Percent Range 98%
Molecular Formula C9 H10 N2 S
CAS Min % 97.5
MDL Number MFCD00005790
Infrared Spectrum Authentic
Packaging Glass bottle