missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile, Thermo Scientific Chemicals

$404.00 - $404.00

Chemical Identifiers

CAS 70291-62-2
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00121495
InChI Key RILAXFOADRDGPQ-UHFFFAOYSA-N
PubChem CID 258182
IUPAC Name 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
SMILES NC1=C(C#N)C2=C(CCC2)S1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3205906
View Documents Promotion Details
Thermo Scientific Chemicals
H3205906
5 g
Each for $404.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70291-62-2
164.23
RILAXFOADRDGPQ-UHFFFAOYSA-N
2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
C8H8N2S
MFCD00121495
258182
NC1=C(C#N)C2=C(CCC2)S1

Specifications

70291-62-2
C8H8N2S
5 g
2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4,5-trimethylene-3-thiophenecarbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-5,6-dihydro, 2-amino-3-cyano-5,6-dihydro-4h-cyclopenta b thiophene, 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile,2-amino-5,6-dihydro, maybridge1_003206, 4,5-trimethylene-2-aminothiophene-3-carbonitrile, 2-amino-5,6-dihydro-4h-cyclopenta b thioph ene-3-carbonitrile
NC1=C(C#N)C2=C(CCC2)S1
164.23
164.23
153°C to 154°C
MFCD00121495
UN3439
RILAXFOADRDGPQ-UHFFFAOYSA-N
2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
258182
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only