missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5,6-dichlorobenzimidazole, 96%, Thermo Scientific™

$167.49 - $167.49

Chemical Identifiers

CAS 18672-03-2
Molecular Formula C7H5Cl2N3
Molecular Weight (g/mol) 202.038
MDL Number MFCD08234602
InChI Key HAADRTMPUSJNOI-UHFFFAOYSA-N
Synonym 2-amino-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzo d imidazol-2-amine, 2-amino-5,6-dichloro-benzimidazole, 2-amino-5,6-dichloro-1h-benzimidazole, 5,6-dichloro-1h-1,3-benzodiazol-2-amine, acmc-209eop, chembl71905, tos-bb-0386, 5,6-dichlorobenzimidazole-2-ylamine, 1h-benzimidazol-2-amine,5,6-dichloro
PubChem CID 466016
IUPAC Name 5,6-dichloro-1H-benzimidazol-2-amine
SMILES C1=C2C(=CC(=C1Cl)Cl)N=C(N2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54597MD
View Documents Promotion Details
Thermo Scientific Chemicals
H54597MD
250 mg
Each for $167.49
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Amino-5,6-dichlorobenzimidazole is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

18672-03-2
202.038
HAADRTMPUSJNOI-UHFFFAOYSA-N
466016
C1=C2C(=CC(=C1Cl)Cl)N=C(N2)N
C7H5Cl2N3
MFCD08234602
2-amino-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzo d imidazol-2-amine, 2-amino-5,6-dichloro-benzimidazole, 2-amino-5,6-dichloro-1h-benzimidazole, 5,6-dichloro-1h-1,3-benzodiazol-2-amine, acmc-209eop, chembl71905, tos-bb-0386, 5,6-dichlorobenzimidazole-2-ylamine, 1h-benzimidazol-2-amine,5,6-dichloro
5,6-dichloro-1H-benzimidazol-2-amine

Specifications

18672-03-2
C7H5Cl2N3
250 mg
HAADRTMPUSJNOI-UHFFFAOYSA-N
5,6-dichloro-1H-benzimidazol-2-amine
466016
96%
244°C to 246°C
MFCD08234602
2-amino-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzo d imidazol-2-amine, 2-amino-5,6-dichloro-benzimidazole, 2-amino-5,6-dichloro-1h-benzimidazole, 5,6-dichloro-1h-1,3-benzodiazol-2-amine, acmc-209eop, chembl71905, tos-bb-0386, 5,6-dichlorobenzimidazole-2-ylamine, 1h-benzimidazol-2-amine,5,6-dichloro
C1=C2C(=CC(=C1Cl)Cl)N=C(N2)N
202.038
202.04
2-Amino-5,6-dichlorobenzimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'