Learn More
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0968303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride | |
186°C to 190°C | |
C7H7ClF3NS | |
1 g | |
4273774 | |
FIAGYDIJZOWVAB-UHFFFAOYSA-N | |
2-amino-4-(trifluoromethyl)benzenethiol;hydrochloride | |
2734678 | |
97% |
4274-38-8 | |
2.46 | |
MFCD00042600 | |
UN1759 | |
2-amino-4-trifluoromethyl benzenethiol hydrochloride, 3-amino-4-mercaptobenzotrifluoride hydrochloride, 2-amino-4-trifluoromethylthiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol hydrochloride, 2-amino-4-trifluoromethyl thiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol, chloride, pubchem2827, c7h6f3ns.hcl, dsstox_cid_31681, dsstox_rid_97532 | |
[H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F | |
229.65 | |
229.65 |
Chemical Identifiers
4274-38-8 | |
229.65 | |
FIAGYDIJZOWVAB-UHFFFAOYSA-N | |
2734678 | |
[H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F |
C7H7ClF3NS | |
MFCD00042600 | |
2-amino-4-trifluoromethyl benzenethiol hydrochloride, 3-amino-4-mercaptobenzotrifluoride hydrochloride, 2-amino-4-trifluoromethylthiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol hydrochloride, 2-amino-4-trifluoromethyl thiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol, chloride, pubchem2827, c7h6f3ns.hcl, dsstox_cid_31681, dsstox_rid_97532 | |
2-amino-4-(trifluoromethyl)benzenethiol;hydrochloride |
Safety and Handling
GHS H Statement
H314-H318-H373-H302-H312-H332-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H312+H332-H314-H317-H335-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 429-560-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only