Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(trifluoromethyl)pyrimidine, 99%

$119.15 - $518.74

Chemical Identifiers

Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyrimidine
CAS 16075-42-6
Molecular Formula C5H4F3N3
Formula Weight 163.1
MDL Number MFCD00828624
Synonym 4-Trifluoromethyl-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5035803

 
alfa aesar™
H5035803
1g Each for $119.15

AAH5035806

 
alfa aesar™
H5035806
5g Each for $518.74
Description & Specifications

Specifications

Formula Weight 163.1
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyrimidine
Assay 99%
Molecular Formula C5H4F3N3
Melting Point 68° to 70°C
MDL Number MFCD00828624
CAS 16075-42-6
Quantity 1g
Synonym 4-Trifluoromethyl-2-pyrimidinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.