missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC442645000

 View more versions of this product

Catalog No. AC442645000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-4-(trifluoromethyl)pyridine
Brown to White
106447-97-6
100.0
C6H5F3N2
RWGBXAQMUBGGKQ-UHFFFAOYSA-N
4-(trifluoromethyl)pyridin-2-amine
2778315
Powder, Solid or Crystalline
Authentic
500mg
96.0
97%
2-amino-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-amine, 4-trifluoromethyl-pyridin-2-ylamine, 2-amino-4-trifluoromethylpyridine, 4-trifluoromethyl-2-pyridinamine, 4-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 4-trifluoromethyl, 4-trifluoromethyl-2-pyridinamine, 2-amino-4-trifluoromethyl-pyridine, pubchem2977
C1=CN=C(C=C1C(F)(F)F)N
162.115
162.11
≥96.0%

Chemical Identifiers

106447-97-6
162.115
2-amino-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-amine, 4-trifluoromethyl-pyridin-2-ylamine, 2-amino-4-trifluoromethylpyridine, 4-trifluoromethyl-2-pyridinamine, 4-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 4-trifluoromethyl, 4-trifluoromethyl-2-pyridinamine, 2-amino-4-trifluoromethyl-pyridine, pubchem2977
4-(trifluoromethyl)pyridin-2-amine
C6H5F3N2
RWGBXAQMUBGGKQ-UHFFFAOYSA-N
2778315
C1=CN=C(C=C1C(F)(F)F)N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents