missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 106447-97-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H5F3N2
Formula Weight 162.11
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442641000 SDS Thermo Fisher Scientific
AC442641000
100mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442645000 SDS Thermo Fisher Scientific
AC442645000
500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

106447-97-6
100.0
C6H5F3N2
2-Amino-4-(trifluoromethyl)pyridine
96.0
97%
162.11
≥96.0%

Specifications

Powder, Solid or Crystalline
100mg
100.0
C6H5F3N2
106447-97-6
2-Amino-4-(trifluoromethyl)pyridine
162.11
White to pale brown
96.0
97%
Authentic
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning