Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-thiazoleacetic acid hydrochloride, 98%

$67.66 - $197.39

Chemical Identifiers

Chemical Name or Material 2-Amino-4-thiazoleacetic acid hydrochloride
CAS 66659-20-9
Molecular Formula C5H6N2O2S·HCl
Formula Weight 194.64
MDL Number MFCD06658943
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2884006

  SDS
alfa aesar™
H2884006
5g Each for $67.66

AAH2884014

  SDS
alfa aesar™
H2884014
25g Each for $197.39
Description & Specifications

Specifications

Formula Weight 194.64
CAS 66659-20-9
Chemical Name or Material 2-Amino-4-thiazoleacetic acid hydrochloride
Melting Point ≈152°C
Molecular Formula C5H6N2O2S·HCl
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 5g
MDL Number MFCD06658943
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.