Please login to your online account to display your discounted pricing

2-Amino-4-tert-butylphenol, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  185830100

 View more versions of this product

Catalog No. AC185830100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 165.23
Quantity 10g
Color Beige to brown
EINECS Number 214-844-4
TSCA TSCA
Melting Point 160° to 164°C
CAS Min % 96.0
CAS Max % 100.0
Linear Formula (CH3)3CC6H3(NH2)OH
MDL Number MFCD00007698
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 2-Amino-4-tert-butylphenol
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H15NO
CAS 1199-46-8
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

Specifications

Chemical Name or Material 2-Amino-4-tert-butylphenol
Name Note 97%
CAS 1199-46-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H15NO
Linear Formula (CH3)3CC6H3(NH2)OH
Formula Weight 165.23
MDL Number MFCD00007698
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents