missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-tert-butylphenol, 97%, ACROS Organics™

$50.84 - $50.84

Chemical Identifiers

CAS 1199-46-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H15NO
Linear Formula (CH3)3CC6H3(NH2)OH
Formula Weight 165.23
MDL Number MFCD00007698
Chemical Name or Material 2-Amino-4-tert-butylphenol, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC185830100 SDS Thermo Fisher Scientific
185830100
Glass bottle 10g Each for $50.84
Specifications Description & Specifications

Chemical Identifiers

1199-46-8
100.0
C10H15NO
165.23
2-Amino-4-tert-butylphenol, 97%
96.0
97%
(CH3)3CC6H3(NH2)OH
MFCD00007698
≥96.0%

Specifications

Crystalline Powder
10g
160° to 164°C
100.0
(CH3)3CC6H3(NH2)OH
C10H15NO
97%
2-Amino-4-tert-butylphenol, 97%
165.23
Beige to brown
96.0
Authentic
MFCD00007698
1199-46-8
Glass bottle
≥96.0%