Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-phenylthiazole hydrobromide monohydrate, 99%

$131.15 - $377.12

Chemical Identifiers

Chemical Name or Material 2-Amino-4-phenylthiazole hydrobromide monohydrate
CAS 52253-69-7
Molecular Formula C9H8N2S·HBr·H2O
Formula Weight 275.17 (257.15 Anhydrous)
MDL Number MFCD00150083
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1954214

 
alfa aesar™
A1954214
25g Each for $131.15

AAA1954222

 
alfa aesar™
A1954222
100g Each for $377.12
Description & Specifications

Specifications

Formula Weight 275.17 (257.15 Anhydrous)
Chemical Name or Material 2-Amino-4-phenylthiazole hydrobromide monohydrate
Melting Point 180° to 183°C
Assay 99%
Quantity 25g
Molecular Formula C9H8N2S·HBr·H2O
MDL Number MFCD00150083
CAS 52253-69-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.