missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-4-phenylthiazole, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 2010-06-2 |
---|---|
Molecular Formula | C9H8N2S |
Molecular Weight (g/mol) | 176.24 |
MDL Number | MFCD00039680 |
InChI Key | PYSJLPAOBIGQPK-UHFFFAOYSA-N |
Synonym | 4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole |
PubChem CID | 40302 |
IUPAC Name | 4-phenyl-1,3-thiazol-2-amine |
SMILES | C1=CC=C(C=C1)C2=CSC(=N2)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC432490010
|
Thermo Scientific Chemicals
432490010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC432490050
|
Thermo Scientific Chemicals
432490050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2010-06-2 | |
176.24 | |
PYSJLPAOBIGQPK-UHFFFAOYSA-N | |
40302 | |
C1=CC=C(C=C1)C2=CSC(=N2)N |
C9H8N2S | |
MFCD00039680 | |
4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole | |
4-phenyl-1,3-thiazol-2-amine |
Specifications
2010-06-2 | |
100.0 | |
96% min. (HPLC) | |
C9H8N2S | |
1g | |
PYSJLPAOBIGQPK-UHFFFAOYSA-N | |
4-phenyl-1,3-thiazol-2-amine | |
40302 | |
97% |
96.0 | |
Authentic | |
Glass bottle | |
MFCD00039680 | |
4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole | |
C1=CC=C(C=C1)C2=CSC(=N2)N | |
176.24 | |
176.24 | |
2-Amino-4-phenylthiazole |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 217-926-8
RUO â Research Use Only