Please login to your online account to display your discounted pricing

2-Amino-4-nitrophenol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-4-nitrophenol
CAS 99-57-0
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H6N2O3
Formula Weight 154.12
MDL Number MFCD00007695
Synonym 2-Hydroxy-5-nitroaniline
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC400892500

  SDS
thermo fisher scientific
AC400892500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC400890250

  SDS
thermo fisher scientific
AC400890250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC400890500

  SDS
fisher scientific acros
400890500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.12
Quantity 250g
Density 100g/mL
Beilstein 13, II, 192
CAS Max % 100.0
Chemical Name or Material 2-Amino-4-nitrophenol
Assay Percent Range 99%
Molecular Formula C6H6N2O3
MDL Number MFCD00007695
CAS 99-57-0
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Synonym 2-Hydroxy-5-nitroaniline
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.