missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-nitrophenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC400890250

 View more versions of this product

Catalog No. AC400890250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

154.12
13, II, 192
100.0
2-Hydroxy-5-nitroaniline
C6H6N2O3
MFCD00007695
Glass bottle
25g
100g/mL
2-Amino-4-nitrophenol
99%
99-57-0
98.5

Chemical Identifiers

99-57-0
100.0
C6H6N2O3
MFCD00007695
2-Amino-4-nitrophenol
98.5
99%
154.12
2-Hydroxy-5-nitroaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.