Learn More
2-Amino-4-nitrobenzoic acid, ≥97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6483122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Amino-4-nitrobenzoic acid | |
257°C (literature) | |
MFCD00007262 | |
785071 | |
UEALKTCRMBVTFN-UHFFFAOYSA-N | |
2-amino-4-nitrobenzoic acid | |
12076 | |
182.13 | |
Solid |
619-17-0 | |
C7H6N2O4 | |
100 g | |
2-amino-4-nitrobenzoic acid, 4-nitroanthranilic acid, benzoic acid, 2-amino-4-nitro, 2-amino-4-nitrobenzoicacid, anthranilic acid, 4-nitro, ccris 441, 2-amino-4-nitro-benzoic acid, 4-nitro-2-aminobenzoic acid, dsstox_cid_963, dsstox_rid_75891 | |
NC1=CC(=CC=C1C(O)=O)[N+]([O-])=O | |
182.14 | |
CHEBI:75335 | |
≥97% |
Chemical Identifiers
619-17-0 | |
182.14 | |
UEALKTCRMBVTFN-UHFFFAOYSA-N | |
12076 | |
2-amino-4-nitrobenzoic acid |
C7H6N2O4 | |
MFCD00007262 | |
2-amino-4-nitrobenzoic acid, 4-nitroanthranilic acid, benzoic acid, 2-amino-4-nitro, 2-amino-4-nitrobenzoicacid, anthranilic acid, 4-nitro, ccris 441, 2-amino-4-nitro-benzoic acid, 4-nitro-2-aminobenzoic acid, dsstox_cid_963, dsstox_rid_75891 | |
CHEBI:75335 | |
NC1=CC(=CC=C1C(O)=O)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 210-583-5
RTECSNumber : CB3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only