Please login to your online account to display your discounted pricing

2-Amino-4-methylthiazole 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  104100250

 View more versions of this product

Catalog No. AC104100250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 114.16
Physical Form Crystalline Solid
Quantity 25g
Color Beige to brown
TSCA TSCA
EINECS Number 216-505-6
Melting Point 42° to 47°C
Beilstein 27, 159
Merck Index 15, 447
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Molecular Formula C4H6N2S
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00005329
CAS 1603-91-4
Chemical Name or Material 2-Amino-4-methylthiazole
Name Note 98%
Assay Percent Range 98%
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material 2-Amino-4-methylthiazole
Name Note 98%
CAS 1603-91-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H6N2S
Formula Weight 114.16
MDL Number MFCD00005329
Accessories

HYGROSCOPIC

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents