Please login to your online account to display your discounted pricing

2-Amino-4-methylthiazole 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-4-methylthiazole
Name Note 98%
CAS 1603-91-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H6N2S
Formula Weight 114.16
MDL Number MFCD00005329
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC104100250

  SDS
fisher scientific acros
104100250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC104101000

  SDS
thermo fisher scientific
104101000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 114.16
Physical Form Crystalline Solid
Quantity 25g
Color Beige to brown
EINECS Number 216-505-6
TSCA TSCA
Melting Point 42° to 47°C
Beilstein 27, 159
Merck Index 15, 447
CAS Max % 100.0
Chemical Name or Material 2-Amino-4-methylthiazole
Molecular Formula C4H6N2S
MDL Number MFCD00005329
CAS 1603-91-4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Assay Percent Range 98%
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 1 HYGROSCOPIC
Packaging Glass bottle
Safety and Handling

HYGROSCOPIC

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.