missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-methylpyrimidine 97%, ACROS Organics™

Manufacturer:  Acros Organics 104091000

 View more versions of this product

Catalog No. AC104091000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-4-methylpyrimidine
Plastic bottle
155°C to 160°C
97%
96.0
97%
MFCD00006101
2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine
CC1=NC(=NC=C1)N
109.132
109.13
≥96.0%
Authentic
White to Brown
100g
108-52-1
100.0
C5H7N3
24, 84
GHCFWKFREBNSPC-UHFFFAOYSA-N
4-methylpyrimidin-2-amine
7939
Crystalline Powder

Chemical Identifiers

108-52-1
109.132
GHCFWKFREBNSPC-UHFFFAOYSA-N
7939
CC1=NC(=NC=C1)N
C5H7N3
MFCD00006101
2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine
4-methylpyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-591-5

RTECSNumber : UV6485000

TSCA : TSCA

SDS Documents