missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54375MD

 View more versions of this product

Catalog No. AAH54375MD



Description

Description

Applications
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester
944401-55-2
MFCD18072554
Slightly soluble in water.
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2C)N
235.094
235.09
250mg
C11H18BN3O2
2-amino-4-methylpyrimidin-5-yl boronic acid pinacol ester
SYJMHOBGFXCKRG-UHFFFAOYSA-N
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
52987906
96%

Chemical Identifiers

944401-55-2
235.094
SYJMHOBGFXCKRG-UHFFFAOYSA-N
52987906
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2C)N
C11H18BN3O2
MFCD18072554
2-amino-4-methylpyrimidin-5-yl boronic acid pinacol ester
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents