Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-methylpyridine, 98%

$27.43 - $27.43

Chemical Identifiers

Chemical Name or Material 2-Amino-4-methylpyridine
CAS 695-34-1
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006321
UN Number UN2811
Beilstein 107066
Merck Index 14,466
Synonym 2-Amino-4-picoline; 4-Methyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1417714

 
alfa aesar™
A1417714
25g Each for $27.43

AAA1417736

 
alfa aesar™
A1417736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.14
Chemical Name or Material 2-Amino-4-methylpyridine
TSCA TSCA
Assay 98%
Molecular Formula C6H8N2
Melting Point 97° to 101°C
Boiling Point 231°C
MDL Number MFCD00006321
CAS 695-34-1
UN Number UN2811
Quantity 25g
Beilstein 107066
Flash Point 118°C (244°F)
Odor Unpleasant
Merck Index 14,466
Synonym 2-Amino-4-picoline; 4-Methyl-2-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
If eye irritation persists: Get medical advice/attention.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number TJ5150000

Applications
2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

Solubility
Soluble in water (41 mg/ml at 20°C), lower alcohols (slightly), chloroform, ethyl acetate, and DMF.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water. Store at -20°C.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H301-H311-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.