missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444530010

 View more versions of this product

Catalog No. AC444530010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Amino-4-methylphenylboronic acid pinacol ester
48°C to 50°C
863578-36-3
C13H20BNO2
PEXRISJTWBBZRY-UHFFFAOYSA-N
17750209
97%
Glass Bottle
1g
96% min. (GC)
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine, 2-amino-4-methylphenylboronic acid pinacol ester, 2-amino-4-methylphenylboronic acid, pinacol ester, 5-methyl-2--4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, benzenamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pexrisjtwbbzry-uhfffaoysa-n
233.12
233.12

Chemical Identifiers

863578-36-3
233.12
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine, 2-amino-4-methylphenylboronic acid pinacol ester, 2-amino-4-methylphenylboronic acid, pinacol ester, 5-methyl-2--4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, benzenamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pexrisjtwbbzry-uhfffaoysa-n
C13H20BNO2
PEXRISJTWBBZRY-UHFFFAOYSA-N
17750209
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents