missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 863578-36-3
Molecular Formula C13H20BNO2
Molecular Weight (g/mol) 233.12
InChI Key PEXRISJTWBBZRY-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine
PubChem CID 17750209
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444530010 SDS View Documents Thermo Fisher Scientific
AC444530010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444530050 SDS View Documents Thermo Fisher Scientific
AC444530050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

863578-36-3
233.12
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine
C13H20BNO2
PEXRISJTWBBZRY-UHFFFAOYSA-N
17750209

Specifications

Glass Bottle
1g
96% min. (GC)
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine
233.12
233.12
2-Amino-4-methylphenylboronic acid pinacol ester
48°C to 50°C
863578-36-3
C13H20BNO2
PEXRISJTWBBZRY-UHFFFAOYSA-N
17750209
97%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning