missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-methylbenzonitrile, 98%

Manufacturer:  Alfa Aesar™ 4326006

 View more versions of this product

Catalog No. AA4326006



Specifications

Specifications

2-Amino-4-methylbenzonitrile
5g
C8H8N2
UN3439
LGNVAEIITHYWCG-UHFFFAOYSA-N
2-amino-4-methylbenzonitrile
2801276
Powder
91°C to 93°C
26830-96-6
MFCD00173706
2-amino-4-methyl-benzonitrile
CC1=CC(=C(C=C1)C#N)N
132.166
132.17
98%

Chemical Identifiers

26830-96-6
132.166
LGNVAEIITHYWCG-UHFFFAOYSA-N
2801276
CC1=CC(=C(C=C1)C#N)N
C8H8N2
MFCD00173706
2-amino-4-methyl-benzonitrile
2-amino-4-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents