missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H53220MD

 View more versions of this product

Catalog No. AAH53220MD



Description

Description

Applications
2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride
250mg
C14H21BClNO4
methyl 3-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid, pinacol ester, hcl, 2-amino-4-methoxycarbonylphenyl boronic acid pinacol ester hydrochloride, methyl 3-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate hydrochloride, methyl 3-amino-4-boronobenzoate, pinacol ester, hcl, 2-amino-4-methoxycarbonyl benzeneboronic acid pinacol ester hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid,pinacol, methyl 3-amino-4-boronobenzoate pinacol ester hydrochloride, 2-amino-4-methoxycarbonyl benzeneboronic acid, pinacol ester hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid pinacol ester hydrochloride
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OC)N.Cl
313.585
313.59
92°C to 94°C
850567-49-6
MFCD06659918
HEBCDUUNEDAXAA-UHFFFAOYSA-N
methyl 3-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate;hydrochloride
44119628
95%

Chemical Identifiers

850567-49-6
313.585
HEBCDUUNEDAXAA-UHFFFAOYSA-N
44119628
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OC)N.Cl
C14H21BClNO4
MFCD06659918
methyl 3-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid, pinacol ester, hcl, 2-amino-4-methoxycarbonylphenyl boronic acid pinacol ester hydrochloride, methyl 3-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate hydrochloride, methyl 3-amino-4-boronobenzoate, pinacol ester, hcl, 2-amino-4-methoxycarbonyl benzeneboronic acid pinacol ester hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid,pinacol, methyl 3-amino-4-boronobenzoate pinacol ester hydrochloride, 2-amino-4-methoxycarbonyl benzeneboronic acid, pinacol ester hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid pinacol ester hydrochloride
methyl 3-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents