missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H5323403

 View more versions of this product

Catalog No. AAH5323403



Specifications

Specifications

2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride
1g
C8H11BClNO4
2-amino-4-methoxycarbonyl phenyl boronic acid hydrochloride, 2-amino-4-methoxycarbonyl phenylboronic acid hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid, hcl, 2-amino-4-methoxycarbonylphenyl boronic acid hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid hydrochloride, 2-amino-4-methoxycarbonyl boronic acid hydrochloride, 2-borono-5-methoxycarbonyl aniline hydrochloride, 4-methoxycarbonyl-2-aminophenylboronic acid hydrochloride, methyl 3-amino-4-boronobenzoate hydrochloride, 2-amino-4-methoxycarbonyl benzeneboronic acid hydrochloride
B(C1=C(C=C(C=C1)C(=O)OC)N)(O)O.Cl
231.439
231.44
200°C to 204°C
380430-55-7
MFCD02258941
IDUSDMZTKZZVAW-UHFFFAOYSA-N
(2-amino-4-methoxycarbonylphenyl)boronic acid;hydrochloride
16427083
95%

Chemical Identifiers

380430-55-7
231.439
IDUSDMZTKZZVAW-UHFFFAOYSA-N
16427083
B(C1=C(C=C(C=C1)C(=O)OC)N)(O)O.Cl
C8H11BClNO4
MFCD02258941
2-amino-4-methoxycarbonyl phenyl boronic acid hydrochloride, 2-amino-4-methoxycarbonyl phenylboronic acid hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid, hcl, 2-amino-4-methoxycarbonylphenyl boronic acid hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid hydrochloride, 2-amino-4-methoxycarbonyl boronic acid hydrochloride, 2-borono-5-methoxycarbonyl aniline hydrochloride, 4-methoxycarbonyl-2-aminophenylboronic acid hydrochloride, methyl 3-amino-4-boronobenzoate hydrochloride, 2-amino-4-methoxycarbonyl benzeneboronic acid hydrochloride
(2-amino-4-methoxycarbonylphenyl)boronic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents