missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-methoxybenzothiazole, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC160350250

 View more versions of this product

Catalog No. AC160350250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-4-methoxybenzothiazole
Glass bottle
153.0°C to 155.0°C
5464-79-9
100.0
C8H8N2OS
2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
COC1=C2C(=CC=C1)SC(=N2)N
180.225
180.23
97%
Authentic
Beige
25g
96.0
97%
MFCD00005792
YEBCRAVYUWNFQT-UHFFFAOYSA-N
4-methoxy-1,3-benzothiazol-2-amine
21622
Crystalline Powder

Chemical Identifiers

5464-79-9
180.225
YEBCRAVYUWNFQT-UHFFFAOYSA-N
21622
COC1=C2C(=CC=C1)SC(=N2)N
C8H8N2OS
MFCD00005792
2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
4-methoxy-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents