Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-methoxy-6-methylpyrimidine, 99%

$53.42 - $169.32

Chemical Identifiers

Chemical Name or Material 2-Amino-4-methoxy-6-methylpyrimidine
CAS 7749-47-5
Molecular Formula C6H9N3O
Formula Weight 139.16
MDL Number MFCD00059768
Beilstein 121733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2467306

  SDS
alfa aesar™
B2467306
5g Each for $53.42

AAB2467314

  SDS
alfa aesar™
B2467314
25g Each for $169.32
Description & Specifications

Specifications

Formula Weight 139.16
Chemical Name or Material 2-Amino-4-methoxy-6-methylpyrimidine
Assay 99%
Molecular Formula C6H9N3O
Melting Point 158° to 164°C
MDL Number MFCD00059768
CAS 7749-47-5
Quantity 5g
Beilstein 121733
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.