missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-hydroxyquinoline hydrate, 97%, water ca 10%

Manufacturer:  Alfa Aesar™ L1276006

 View more versions of this product

Catalog No. AAL1276006



Specifications

Specifications

2-Amino-4-hydroxyquinoline hydrate
5g
water ≈10%
MFCD00052386
2-aminoquinolin-4-ol, 4-quinolinol, 2-amino, 2-amino-4-hydroxyquinoline, 2-aminoquinolin-4 1h-one, 2-amino-4-1h-quinolinone, 2-amino-quinolin-4-ol, 2-amino-4 1h-quinolinone, 4 1h-quinolinone, 2-amino, 2-?aminoquinolin-?4-?ol, aminoquinolinol
C1=CC=C2C(=C1)C(=O)C=C(N2)N
160.176
160.18 (Anhydrous)
~298°C
42712-64-1
C9H8N2O
124785
LWGUCIXHBVVATR-UHFFFAOYSA-N
2-amino-1H-quinolin-4-one
594793
97%

Chemical Identifiers

42712-64-1
160.176
LWGUCIXHBVVATR-UHFFFAOYSA-N
594793
C1=CC=C2C(=C1)C(=O)C=C(N2)N
C9H8N2O
MFCD00052386
2-aminoquinolin-4-ol, 4-quinolinol, 2-amino, 2-amino-4-hydroxyquinoline, 2-aminoquinolin-4 1h-one, 2-amino-4-1h-quinolinone, 2-amino-quinolin-4-ol, 2-amino-4 1h-quinolinone, 4 1h-quinolinone, 2-amino, 2-?aminoquinolin-?4-?ol, aminoquinolinol
2-amino-1H-quinolin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents