Learn More
2-Amino-4-hydroxypyridine, 98%, Thermo Scientific Chemicals
$66.79 - $276.16
Chemical Identifiers
CAS | 33631-05-9 |
---|---|
Molecular Formula | C5H6N2O |
Molecular Weight (g/mol) | 110.116 |
MDL Number | MFCD04114161 |
InChI Key | HQNIMNQVKVPZES-UHFFFAOYSA-N |
Synonym | 2-amino-4-hydroxypyridine, 2-aminopyridin-4-ol, 2-amino-4-pyridinol, 4-pyridinol, 2-amino, 2-amino-pyridin-4-ol, zlchem 31, pubchem6677, acmc-1acz5, 4-hydroxypyridine-2-amine, 2-aminopyridin-4 1h-one |
PubChem CID | 820936 |
IUPAC Name | 2-amino-1H-pyridin-4-one |
SMILES | C1=CNC(=CC1=O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6483603
|
Thermo Scientific Chemicals
H6483603 |
1 g |
Each for $66.79
|
|
|||||
AAH6483606
|
Thermo Scientific Chemicals
H6483606 |
5 g |
Each for $276.16
|
|
|||||
Description
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
33631-05-9 | |
110.116 | |
HQNIMNQVKVPZES-UHFFFAOYSA-N | |
820936 | |
C1=CNC(=CC1=O)N |
C5H6N2O | |
MFCD04114161 | |
2-amino-4-hydroxypyridine, 2-aminopyridin-4-ol, 2-amino-4-pyridinol, 4-pyridinol, 2-amino, 2-amino-pyridin-4-ol, zlchem 31, pubchem6677, acmc-1acz5, 4-hydroxypyridine-2-amine, 2-aminopyridin-4 1h-one | |
2-amino-1H-pyridin-4-one |
Specifications
33631-05-9 | |
C5H6N2O | |
1 g | |
2-amino-4-hydroxypyridine, 2-aminopyridin-4-ol, 2-amino-4-pyridinol, 4-pyridinol, 2-amino, 2-amino-pyridin-4-ol, zlchem 31, pubchem6677, acmc-1acz5, 4-hydroxypyridine-2-amine, 2-aminopyridin-4 1h-one | |
HQNIMNQVKVPZES-UHFFFAOYSA-N | |
2-amino-1H-pyridin-4-one | |
820936 | |
98% | |
2-Amino-4-hydroxypyridine |
Yellow | |
MFCD04114161 | |
385930 | |
Slightly soluble in water. | |
C1=CNC(=CC1=O)N | |
110.116 | |
110.12 | |
Solid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only