missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-hydroxybenzothiazole 98.0+%, TCI America™

$198.58 - $591.03

Chemical Identifiers

CAS 7471-03-6
Molecular Formula C7H6N2OS
Molecular Weight (g/mol) 166.20
MDL Number MFCD00160060
InChI Key PFQJPSASUCHKRO-UHFFFAOYSA-N
Synonym 2-Aminobenzo[d]thiazol-4-ol
PubChem CID 24057
IUPAC Name 2-amino-1,3-benzothiazol-4-ol
SMILES C1=CC(=C2C(=C1)SC(=N2)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A30011G
View Documents
Tci America
A30011G
1g
Each for $198.58
 
A30015G
View Documents
Tci America
A30015G
5g
Each for $591.03
 
Description

Description

Specifications

Chemical Identifiers

7471-03-6
166.20
PFQJPSASUCHKRO-UHFFFAOYSA-N
24057
C1=CC(=C2C(=C1)SC(=N2)N)O
C7H6N2OS
MFCD00160060
2-Aminobenzo[d]thiazol-4-ol
2-amino-1,3-benzothiazol-4-ol

Specifications

Undesignated
1g
C7H6N2OS
2-Aminobenzo[d]thiazol-4-ol
C1=CC(=C2C(=C1)SC(=N2)N)O
166.20
166.20
≥98.0% (T)
184°C
7471-03-6
MFCD00160060
PFQJPSASUCHKRO-UHFFFAOYSA-N
2-amino-1,3-benzothiazol-4-ol
24057
Crystal-Powder at 20°C
2-Amino-4-hydroxybenzothiazole
Safety and Handling

Safety and Handling

RTECSNumber : DL1800000

TSCA : No

SDS