Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-hydroxy-5,6-dimethylpyrimidine, 97%

$82.20 - $298.62

Chemical Identifiers

Chemical Name or Material 2-Amino-4-hydroxy-5,6-dimethylpyrimidine
CAS 3977-23-9
Molecular Formula C6H9N3O
Formula Weight 139.16
MDL Number MFCD00023258
Synonym 2-Amino-5,6-dimethyl-4-hydroxypyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4343203

 
alfa aesar™
4343203
1g Each for $82.20

AA4343206

 
alfa aesar™
4343206
5g Each for $298.62
Description & Specifications

Specifications

Formula Weight 139.16
Chemical Name or Material 2-Amino-4-hydroxy-5,6-dimethylpyrimidine
Color White
Melting Point 316° to 318°C (dec.)
Physical Form Powder
Assay 97%
Quantity 1g
Molecular Formula C6H9N3O
MDL Number MFCD00023258
CAS 3977-23-9
Synonym 2-Amino-5,6-dimethyl-4-hydroxypyrimidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.