missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-hydroxy-5,6-dimethylpyrimidine, 97%

Manufacturer:  Alfa Aesar™ 4343214

 View more versions of this product

Catalog No. AA4343214



Specifications

Specifications

2-Amino-4-hydroxy-5,6-dimethylpyrimidine
316°C to 318°C (decomposition)
3977-23-9
MFCD00023258
APWRLAZEMYLHKZ-UHFFFAOYSA-N
2-amino-5,6-dimethyl-1H-pyrimidin-4-one
77600
Powder
White
25g
C6H9N3O
2-amino-5,6-dimethylpyrimidin-4-ol, 2-amino-5,6-dimethyl-4-hydroxypyrimidine, 2-amino-5,6-dimethylpyrimidin-4 3h-one, 2-amino-5,6-dimethyl-4-pyrimidinol, 2-amino-5,6-dimethyl-4 3h-pyrimidinone, 2-amino-5,6-dimethylpyrimidin-4 1h-one, 2-amino-5,6-dimethyl-3h-pyrimidin-4-one, aminodimethylpyrimidinol, acmc-1cpts, 4 1h-pyrimidinone, 2-amino-5,6-dimethyl
CC1=C(NC(=NC1=O)N)C
139.158
139.16
97%

Chemical Identifiers

3977-23-9
139.158
APWRLAZEMYLHKZ-UHFFFAOYSA-N
77600
CC1=C(NC(=NC1=O)N)C
C6H9N3O
MFCD00023258
2-amino-5,6-dimethylpyrimidin-4-ol, 2-amino-5,6-dimethyl-4-hydroxypyrimidine, 2-amino-5,6-dimethylpyrimidin-4 3h-one, 2-amino-5,6-dimethyl-4-pyrimidinol, 2-amino-5,6-dimethyl-4 3h-pyrimidinone, 2-amino-5,6-dimethylpyrimidin-4 1h-one, 2-amino-5,6-dimethyl-3h-pyrimidin-4-one, aminodimethylpyrimidinol, acmc-1cpts, 4 1h-pyrimidinone, 2-amino-5,6-dimethyl
2-amino-5,6-dimethyl-1H-pyrimidin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents