missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-fluoropyridine, 98%

$55.05 - $724.80

Chemical Identifiers

CAS 944401-77-8
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD09260906
InChI Key WCCFLOQQACDOAX-UHFFFAOYSA-N
Synonym 2-amino-4-fluoropyridine, 4-fluoro-2-pyridinamine, 4-fluoro-2-pyridylamine, 4-fluoro-pyridin-2-ylamine, 4-fluoro-2-pyridineamine, amino-4-fluoropyridine, pubchem19911, acmc-209rrv, 2-pyridinamine,4-fluoro, 4-fluoro-pyridin-2-amine
PubChem CID 18327808
IUPAC Name 4-fluoropyridin-2-amine
SMILES C1=CN=C(C=C1F)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64233MD View Documents Alfa Aesar™
H64233MD
250mg Each for $55.05
 
AAH6423303 View Documents Alfa Aesar™
H6423303
1g Each for $174.26
 
AAH6423306 View Documents Alfa Aesar™
H6423306
5g Each for $724.80
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

944401-77-8
112.107
WCCFLOQQACDOAX-UHFFFAOYSA-N
18327808
C1=CN=C(C=C1F)N
C5H5FN2
MFCD09260906
2-amino-4-fluoropyridine, 4-fluoro-2-pyridinamine, 4-fluoro-2-pyridylamine, 4-fluoro-pyridin-2-ylamine, 4-fluoro-2-pyridineamine, amino-4-fluoropyridine, pubchem19911, acmc-209rrv, 2-pyridinamine,4-fluoro, 4-fluoro-pyridin-2-amine
4-fluoropyridin-2-amine

Specifications

Yellow
944401-77-8
MFCD09260906
2-amino-4-fluoropyridine, 4-fluoro-2-pyridinamine, 4-fluoro-2-pyridylamine, 4-fluoro-pyridin-2-ylamine, 4-fluoro-2-pyridineamine, amino-4-fluoropyridine, pubchem19911, acmc-209rrv, 2-pyridinamine,4-fluoro, 4-fluoro-pyridin-2-amine
C1=CN=C(C=C1F)N
112.107
112.04
98%
250mg
C5H5FN2
15519014
WCCFLOQQACDOAX-UHFFFAOYSA-N
4-fluoropyridin-2-amine
18327808
Solid
2-Amino-4-fluoropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C