missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-fluorophenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2616503

 View more versions of this product

Catalog No. AAH2616503



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-4-fluorophenol
138°C to 141°C
MFCD00077451
5-fluoro-2-hydroxyaniline, phenol, 2-amino-4-fluoro, 2-amino-4-fluoro-phenol, 4-fluoro-2-aminophenol, zlchem 493, pubchem2825, acmc-1ah2l, phenol,2-amino-4-fluoro, 2-amino-4-fluorophenol
C1=CC(=C(C=C1F)N)O
127.118
127.12
399-97-3
C6H6FNO
1 g
ULDFRPKVIZMKJG-UHFFFAOYSA-N
2-amino-4-fluorophenol
2735917
97%

Chemical Identifiers

399-97-3
127.118
ULDFRPKVIZMKJG-UHFFFAOYSA-N
2-amino-4-fluorophenol
C6H6FNO
MFCD00077451
2735917
C1=CC(=C(C=C1F)N)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only