missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chloropyrimidine, 98%

Manufacturer:  Alfa Aesar™ H5500706

 View more versions of this product

Catalog No. AAH5500706



Description

Description

Applications
2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and acids.
Specifications

Specifications

2-Amino-4-chloropyrimidine
5g
C4H4ClN3
2-amine-4-chloropyrimidine
C1=CN=C(N=C1Cl)N
129.547
129.55
155°C to 160°C
3993-78-0
MFCD00038021
DBGFGNCFYUNXLD-UHFFFAOYSA-N
4-chloropyrimidin-2-amine
223332
98%

Chemical Identifiers

3993-78-0
129.547
DBGFGNCFYUNXLD-UHFFFAOYSA-N
223332
C1=CN=C(N=C1Cl)N
C4H4ClN3
MFCD00038021
2-amine-4-chloropyrimidine
4-chloropyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Documents