Learn More
2-Amino-4-chloropyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5500703
Description
2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and acids.
Specifications
2-Amino-4-chloropyrimidine | |
155°C to 160°C (decomposition) | |
MFCD00038021 | |
2-amino-4-chloropyrimidine, 2-amine-4-chloropyrimidine, 2-pyrimidinamine, 4-chloro, 4-chloro-2-pyrimidinamine, 4-chloro-2-aminopyrimidine, 4-chloropyrimidine-2-amine, 4-chloro-pyrimidin-2-ylamine, 4-chloropyrimidine-2-ylamine, 2-amino-6-chloropyrimidine, 4-chloro-2-pyrimidineamine | |
DBGFGNCFYUNXLD-UHFFFAOYSA-N | |
4-chloropyrimidin-2-amine | |
223332 | |
98% |
3993-78-0 | |
C4H4ClN3 | |
1 g | |
Soluble in dimethyl sulfoxide. | |
C1=CN=C(N=C1Cl)N | |
129.547 | |
129.55 |
Chemical Identifiers
3993-78-0 | |
129.547 | |
DBGFGNCFYUNXLD-UHFFFAOYSA-N | |
223332 | |
C1=CN=C(N=C1Cl)N |
C4H4ClN3 | |
MFCD00038021 | |
2-amino-4-chloropyrimidine, 2-amine-4-chloropyrimidine, 2-pyrimidinamine, 4-chloro, 4-chloro-2-pyrimidinamine, 4-chloro-2-aminopyrimidine, 4-chloropyrimidine-2-amine, 4-chloro-pyrimidin-2-ylamine, 4-chloropyrimidine-2-ylamine, 2-amino-6-chloropyrimidine, 4-chloro-2-pyrimidineamine | |
4-chloropyrimidin-2-amine |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only