Learn More
2-Amino-4-chloropyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 441460050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Amino-4-chloropyridine | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD04113820 | |
2-amino-4-chloropyridine, 4-chloro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro, 4-chloro-2-pyridinamine, 4-chloro-2-pyridylamine, 2-amino-4-chloro pyridine, 4-chloro-2-pyridinylamine, 2-amino-4-chloro-pyridine, 4-chloro-2-aminopyridine, 4-chlorpyridin-2-amin | |
NC1=CC(Cl)=CC=[NH+]1 | |
129.57 | |
128.56 |
19798-80-2 | |
100.0 | |
96% min. (GC) | |
C5H6ClN2 | |
5g | |
RQMWVVBHJMUJNZ-UHFFFAOYSA-O | |
4-chloropyridin-2-amine | |
581866 | |
97% |
Chemical Identifiers
19798-80-2 | |
129.57 | |
RQMWVVBHJMUJNZ-UHFFFAOYSA-O | |
581866 | |
NC1=CC(Cl)=CC=[NH+]1 |
C5H6ClN2 | |
MFCD04113820 | |
2-amino-4-chloropyridine, 4-chloro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro, 4-chloro-2-pyridinamine, 4-chloro-2-pyridylamine, 2-amino-4-chloro pyridine, 4-chloro-2-pyridinylamine, 2-amino-4-chloro-pyridine, 4-chloro-2-aminopyridine, 4-chlorpyridin-2-amin | |
4-chloropyridin-2-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
RUO â Research Use Only