Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chlorophenol, 98%

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chlorophenol
CAS 95-85-2
Molecular Formula C6H6ClNO
Formula Weight 143.57
MDL Number MFCD00007694
UN Number UN2673
Beilstein 774859
Synonym 5-Chloro-2-hydroxyaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1013522

  SDS
alfa aesar™
A1013522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1013536

  SDS
alfa aesar™
A1013536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.57
Quantity 100g
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-AMINO-4-CHLOROPHENOL
RTECS Number SJ5700000
Chemical Name or Material 2-Amino-4-chlorophenol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 95-85-2
UN Number UN2673
Melting Point 138° to 142°C
Health Hazard 1 Warning
Beilstein 774859
MDL Number MFCD00007694
Synonym 5-Chloro-2-hydroxyaniline
TSCA TSCA
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C6H6ClNO
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.