Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chlorobenzothiazole hydrobromide, 97%

$62.23 - $62.23

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chlorobenzothiazole hydrobromide
CAS 27058-83-9
Molecular Formula C7H5ClN2S·HBr
Formula Weight 265.57
MDL Number MFCD00035186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0544006

  SDS
alfa aesar™
L0544006
5g Each for $62.23
Description & Specifications

Specifications

Formula Weight 265.57
CAS 27058-83-9
Chemical Name or Material 2-Amino-4-chlorobenzothiazole hydrobromide
Health Hazard 1 Warning
Molecular Formula C7H5ClN2S·HBr
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
RTECS Number DL1670000
Melting Point 240° to 242°C
MDL Number MFCD00035186
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.