Please login to your online account to display your discounted pricing

2-Amino-4-chlorobenzothiazole 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  158470250

 View more versions of this product

Catalog No. AC158470250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Color White to light beige or grayish
Physical Form Crystalline Powder
Formula Weight 184.64
Quantity 25g
EINECS Number 243-439-5
TSCA TSCA
Melting Point 203° to 208°C
RTECS Number DL1225000
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-4-chlorobenzothiazole
Name Note 97%
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C7H5ClN2S
MDL Number MFCD00005791
CAS 19952-47-7
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Amino-4-chlorobenzothiazole
Name Note 97%
CAS 19952-47-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H5ClN2S
Formula Weight 184.64
MDL Number MFCD00005791