missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-chlorobenzothiazole 97%, ACROS Organics™

Manufacturer:  Acros Organics 158470050

 View more versions of this product

Catalog No. AC158470050



Specifications

Specifications

2-Amino-4-chlorobenzothiazole
Glass bottle
203.0°C to 208.0°C
97%
96.0
97%
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
4-chloro-1,3-benzothiazol-2-amine
29872
Crystalline Powder
Authentic
Beige to White or Gray
5g
19952-47-7
100.0
C7H5ClN2S
2-amino-4-chlorobenzothiazole, 2-benzothiazolamine, 4-chloro, 4-chlorobenzo d thiazol-2-amine, 4-chlorobenzothiazol-2-ylamine, benzothiazole, 2-amino-4-chloro, 4-chloro-2-benzothiazolamine, 4-chloro-2-aminobenzothiazole, 4-chloro-benzothiazol-2-ylamine, 4-chlorobenzothiazole-2-ylamine, zlchem 166
C1=CC2=C(C(=C1)Cl)N=C(S2)N
184.641
184.64
97%

Chemical Identifiers

19952-47-7
184.641
OEQQFQXMCPMEIH-UHFFFAOYSA-N
29872
C1=CC2=C(C(=C1)Cl)N=C(S2)N
C7H5ClN2S
MFCD00005791
2-amino-4-chlorobenzothiazole, 2-benzothiazolamine, 4-chloro, 4-chlorobenzo d thiazol-2-amine, 4-chlorobenzothiazol-2-ylamine, benzothiazole, 2-amino-4-chloro, 4-chloro-2-benzothiazolamine, 4-chloro-2-aminobenzothiazole, 4-chloro-benzothiazol-2-ylamine, 4-chlorobenzothiazole-2-ylamine, zlchem 166
4-chloro-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

EINECSNumber : 243-439-5

RTECSNumber : DL1225000

TSCA : TSCA

SDS Documents