Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chlorobenzothiazole, 97%

$78.04 - $664.49

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chlorobenzothiazole
CAS 19952-47-7
Molecular Formula C7H5ClN2S
Formula Weight 184.65
MDL Number MFCD00005791
Beilstein 136784
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1816006

  SDS
alfa aesar™
A1816006
5g Each for $78.04

AAA1816014

  SDS
alfa aesar™
A1816014
25g Each for $242.59

AAA1816022

  SDS
alfa aesar™
A1816022
100g Each for $664.49
Description & Specifications

Specifications

Formula Weight 184.65
Health Hazard 1 Warning
Assay 97%
MDL Number MFCD00005791
Quantity 5g
Molecular Formula C7H5ClN2S
CAS 19952-47-7
Chemical Name or Material 2-Amino-4-chlorobenzothiazole
Melting Point 203° to 205°C
Beilstein 136784
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DL1225000
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.