Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chlorobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chlorobenzonitrile
CAS 38487-86-4
Molecular Formula C7H5ClN2
Formula Weight 152.58
MDL Number MFCD00035927
UN Number UN3439
Synonym 4-Chloroanthranilonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2512009

  SDS
alfa aesar™
B2512009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2512018

  SDS
alfa aesar™
B2512018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.58
Chemical Name or Material 2-Amino-4-chlorobenzonitrile
TSCA TSCA
Melting Point 159° to 162°C
Quantity 10g
Assay 97%
Molecular Formula C7H5ClN2
MDL Number MFCD00035927
CAS 38487-86-4
UN Number UN3439
Synonym 4-Chloroanthranilonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.