Please login to your online account to display your discounted pricing

2-Amino-4-chlorobenzoic acid, 98%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC103325000

 View more versions of this product

Catalog No. AC103325000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (HPLC)
Physical Form Powder
Formula Weight 171.58
Quantity 500g
Color Beige to light brown
Melting Point 231° to 235°C
Beilstein 14, 365
CAS Max % 100.0
Assay Percent Range 98%
Chemical Name or Material 2-Amino-4-chlorobenzoic acid
Molecular Formula C7H6ClNO2
CAS 89-77-0
MDL Number MFCD00007778
CAS Min % 97.5
Synonym 4-chloroanthranilic acid
Linear Formula H2NC6H3(Cl)CO2H
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Amino-4-chlorobenzoic acid
CAS 89-77-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C7H6ClNO2
Linear Formula H2NC6H3(Cl)CO2H
Formula Weight 171.58
MDL Number MFCD00007778
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents